Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL30
Entrez Gene ID
101926789
Gene Name
mitochondrial ribosomal protein L30
Gene Aliases
L30mt, MRP-L30
Chromosome Location
Chr.13: 3462060 - 3489399 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284082.1 NP_001271011.1 4 - 5 351 97
XM_005575061.2 XP_005575118.1 5 - 6 531 97
XM_015433185.1 XP_015288671.1 5 - 6 557 97
XM_015433186.1 XP_015288672.1 4 - 5 317 97
GenBank mRNA AB169083.1 - 4 - 5 351 97

View Full Product Details