Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF3
Entrez Gene ID
102134212
Gene Name
fibroblast growth factor 3
Gene Aliases
-
Chromosome Location
Chr.14: 4928585 - 4937931 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576979.1 XP_005577036.1 2 - 3 812 98

View Full Product Details