Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFNW1
Entrez Gene ID
102114910
Gene Name
interferon omega 1
Gene Aliases
-
Chromosome Location
Chr.15: 57588433 - 57589728 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581634.2 XP_005581691.1 1 - 1 568 96

View Full Product Details