Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC14
Entrez Gene ID
102134314
Gene Name
sialic acid binding Ig like lectin 14
Gene Aliases
SIGLEC5
Chromosome Location
Chr.19: 52342296 - 52379726 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590133.2 XP_005590190.1 4 - 5 911 66

View Full Product Details