Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TREML2
Entrez Gene ID
102131387
Gene Name
triggering receptor expressed on myeloid cells like 2
Gene Aliases
-
Chromosome Location
Chr.4: 129486761 - 129499495 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553058.2 XP_005553115.1 4 - 5 2350 75

View Full Product Details