Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR17
Entrez Gene ID
102128937
Gene Name
G protein-coupled receptor 17
Gene Aliases
-
Chromosome Location
Chr.12: 3973199 - 3980841 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572854.2 XP_005572911.1 1 - 2 287 80

View Full Product Details