Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102116690
Entrez Gene ID
102116690
Gene Name
disintegrin and metalloproteinase domain-containing protein 20
Gene Aliases
ADAM20
Chromosome Location
Chr.7: 135170977 - 135232299 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561647.2 XP_005561704.1 1 - 2 3009 97

View Full Product Details