Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAKMIP3
Entrez Gene ID
102117561
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.9: 131496823 - 131615728 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566756.2 XP_005566813.1 4 - 5 3527 68
XM_005566758.2 XP_005566815.1 4 - 5 1050 68
XM_005566760.2 XP_005566817.1 4 - 5 3527 68
XM_015456882.1 XP_015312368.1 3 - 4 3255 68

View Full Product Details