Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOX10
Entrez Gene ID
102119321
Gene Name
SRY-box transcription factor 10
Gene Aliases
-
Chromosome Location
Chr.10: 13225741 - 13238153 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567327.2 XP_005567384.1 3 - 4 920 96

View Full Product Details