Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JRK
Entrez Gene ID
102131182
Gene Name
Jrk helix-turn-helix protein
Gene Aliases
-
Chromosome Location
Chr.8: 144159016 - 144185275 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015455283.1 XP_015310769.1 1 - 2 3175 69

View Full Product Details