Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD18
Entrez Gene ID
102121933
Gene Name
caspase recruitment domain family member 18
Gene Aliases
-
Chromosome Location
Chr.14: 101099034 - 101101125 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579498.2 XP_005579555.1 2 - 3 329 78

View Full Product Details