Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RP1L1
Entrez Gene ID
102140429
Gene Name
RP1 like 1
Gene Aliases
-
Chromosome Location
Chr.8: 10101074 - 10120096 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562594.2 XP_005562651.1 1 - 2 625 87

View Full Product Details