Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRE1
Entrez Gene ID
102140219
Gene Name
adhesion G protein-coupled receptor E1
Gene Aliases
EMR1
Chromosome Location
Chr.19: 7110323 - 7158119 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015440864.1 XP_015296350.1 14 - 15 2100 63

View Full Product Details