Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102139474
Entrez Gene ID
102139474
Gene Name
kallikrein-2
Gene Aliases
KLK2
Chromosome Location
Chr.19: 51713573 - 51720673 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590049.1 XP_005590106.1 3 - 4 521 90
XM_005590050.1 XP_005590107.1 3 - 4 607 90
XM_005590051.1 XP_005590108.1 3 - 4 521 90
XM_005590052.1 XP_005590109.1 2 - 3 361 90

View Full Product Details