Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102120633
Entrez Gene ID
102120633
Gene Name
protocadherin gamma-C4
Gene Aliases
PCDHGA10
Chromosome Location
Chr.6: 140449187 - 140641638 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015452042.1 XP_015307528.1 1 - 1 1024 115

View Full Product Details