Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK7
Entrez Gene ID
102133307
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
-
Chromosome Location
Chr.14: 8889345 - 8893512 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577264.2 XP_005577321.1 1 - 2 1493 70

View Full Product Details