Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A3GALT2
Entrez Gene ID
102136396
Gene Name
alpha 1,3-galactosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.1: 194630652 - 194644767 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544050.2 XP_005544107.1 2 - 3 1671 82

View Full Product Details