Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLRF2
Entrez Gene ID
102142326
Gene Name
killer cell lectin like receptor F2
Gene Aliases
-
Chromosome Location
Chr.11: 10222208 - 10239110 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570091.2 XP_005570148.1 5 - 6 4952 85
XM_005570092.2 XP_005570149.1 4 - 5 4895 85
XM_005570093.2 XP_005570150.1 5 - 6 730 85
XM_005570094.2 XP_005570151.1 5 - 6 4952 85

View Full Product Details