Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCGB3A1
Entrez Gene ID
102122554
Gene Name
secretoglobin family 3A member 1
Gene Aliases
-
Chromosome Location
Chr.6: 180618832 - 180621248 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558810.2 XP_005558867.1 3 - 4 1231 97

View Full Product Details