Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNMA6A
Entrez Gene ID
102143788
Gene Name
PNMA family member 6A
Gene Aliases
-
Chromosome Location
Chr.X: 150048523 - 150050052 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596082.2 XP_005596139.1 1 - 1 722 84

View Full Product Details