Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPPB
Entrez Gene ID
102115953
Gene Name
natriuretic peptide B
Gene Aliases
-
Chromosome Location
Chr.1: 216258003 - 216260377 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544772.2 XP_005544829.1 1 - 2 1139 72

View Full Product Details