Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAND1
Entrez Gene ID
102132552
Gene Name
heart and neural crest derivatives expressed 1
Gene Aliases
-
Chromosome Location
Chr.6: 153827693 - 153831003 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558356.2 XP_005558413.1 1 - 2 829 63

View Full Product Details