Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB39
Entrez Gene ID
102145777
Gene Name
zinc finger and BTB domain containing 39
Gene Aliases
-
Chromosome Location
Chr.11: 55950350 - 55957681 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571272.2 XP_005571329.1 1 - 1 3323 67

View Full Product Details