Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYPC
Entrez Gene ID
102142616
Gene Name
glycophorin C (Gerbich blood group)
Gene Aliases
-
Chromosome Location
Chr.12: 4979001 - 5024442 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572887.2 XP_005572944.1 4 - 5 711 60
XM_005572888.2 XP_005572945.1 3 - 4 595 60
XM_015431948.1 XP_015287434.1 4 - 5 665 60
XM_015431949.1 XP_015287435.1 4 - 5 675 60
XM_015431950.1 XP_015287436.1 4 - 5 627 60
XM_015431951.1 XP_015287437.1 3 - 4 608 60
XM_015431953.1 XP_015287439.1 3 - 4 560 60
XM_015431954.1 XP_015287440.1 3 - 4 562 60

View Full Product Details