Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102147021
Entrez Gene ID
102147021
Gene Name
chymotrypsinogen B2
Gene Aliases
CTRB1
Chromosome Location
Chr.20: 63385548 - 63388625 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015442750.1 XP_015298236.1 1 - 2 53 107

View Full Product Details