Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CES1
Entrez Gene ID
102129184
Gene Name
carboxylesterase 1
Gene Aliases
-
Chromosome Location
Chr.20: 43746903 - 43792494 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283294.1 NP_001270223.1 7 - 8 902 92
XM_015442712.1 XP_015298198.1 7 - 8 1176 92
GenBank mRNA AB010633.1 - 7 - 8 902 92

View Full Product Details