Product Details

Assay Reference Genome
Location

Chr.7:141191481 on build GRCm38
Cytoband
7F5
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
15461
Gene Symbol
Hras
Gene Name
Harvey rat sarcoma virus oncogene
Gene Aliases
H-ras, Ha-ras, Harvey-ras, Hras-1, Hras1, Kras2, c-H-ras, c-Ha-ras, c-rasHa, ras
Location
Chr.7:141189934-141194004 on build GRCm38
Assay Gene Location
Overlaps Intron 5 - Exon 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Hras NM_001130443.1 NP_001123915.1
NM_001130444.1 NP_001123916.1
NM_008284.2 NP_032310.2
AJ437023.1 CAD24593.1
AY373386.1 AAQ81319.1
BU924724.1
CK788792.1

View Full Product Details