The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007557.3 | NP_031583.2 | 6 - 7 | 1392 | 58 |
GenBank mRNA | BC010771.1 | - | 6 - 7 | 1364 | 58 |
CA316109.1 | - | 4 - 5 | 526 | 58 | |
X56906.1 | - | 6 - 7 | 1243 | 58 |