Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Camk2b
Entrez Gene ID
12323
Gene Name
calcium/calmodulin-dependent protein kinase II, beta
Gene Aliases
-
Chromosome Location
Chr.11: 5969642 - 6065748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174054.1 NP_001167525.1 17 - 18 1402 67
NM_007595.5 NP_031621.3 17 - 18 1441 67
GenBank mRNA AK046454.1 - 17 - 18 1442 67
AK158794.1 - 12 - 13 1085 67
AK163698.1 - 17 - 18 1258 67
BC024803.1 - 17 - 18 1218 67
X63615.1 - 17 - 18 1256 67

View Full Product Details