The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007939.2 | NP_031965.2 | 13 - 14 | 2455 | 73 |
GenBank mRNA | AK220393.1 | - | 11 - 12 | 1939 | 73 |
U72207.1 | - | 13 - 14 | 2464 | 73 |