The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123382.1 | NP_001116854.1 | 10 - 11 | 1611 | 106 |
NM_008362.2 | NP_032388.1 | 10 - 11 | 1542 | 106 | |
GenBank mRNA | AK048550.1 | - | 10 - 11 | 1529 | 106 |
AK054137.1 | - | 10 - 11 | 1612 | 106 | |
BC109135.2 | - | 10 - 11 | 1448 | 106 | |
M20658.1 | - | 10 - 11 | 1542 | 106 |