The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008552.4 | NP_032578.2 | 9 - 9 | 1594 | 96 |
GenBank mRNA | AK015036.1 | - | 9 - 9 | 1585 | 96 |
AK030261.1 | - | 9 - 9 | 1584 | 96 | |
AK030265.1 | - | 9 - 9 | 1595 | 96 | |
BC120790.1 | - | 4 - 4 | 916 | 96 | |
BC125537.1 | - | 5 - 5 | 979 | 96 | |
BC144900.1 | - | 4 - 4 | 928 | 96 |