The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271716.1 | NP_001258645.1 | 4 - 4 | 815 | 78 |
NM_001271717.1 | NP_001258646.1 | 3 - 3 | 727 | 78 | |
NM_001301372.1 | NP_001288301.1 | 5 - 5 | 901 | 78 | |
NM_008560.3 | NP_032586.1 | 3 - 3 | 758 | 78 | |
GenBank mRNA | AK033100.1 | - | 3 - 3 | 697 | 78 |
BC139210.1 | - | 2 - 2 | 661 | 78 | |
BC139211.1 | - | 2 - 2 | 660 | 78 |