The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159520.1 | NP_001152992.1 | 5 - 6 | 1181 | 57 |
NM_008781.4 | NP_032807.3 | 5 - 6 | 1181 | 57 | |
GenBank mRNA | AK012493.1 | - | 5 - 6 | 883 | 57 |
AK014337.1 | - | 5 - 6 | 883 | 57 | |
AK044985.1 | - | 5 - 6 | 886 | 57 | |
AK045018.1 | - | 5 - 6 | 1182 | 57 | |
AK144973.1 | - | 5 - 6 | 886 | 57 | |
AK147919.1 | - | 5 - 6 | 886 | 57 | |
AK148354.1 | - | 5 - 6 | 1162 | 57 | |
AK157760.1 | - | 5 - 6 | 883 | 57 | |
BC048699.1 | - | 5 - 6 | 852 | 57 | |
X59358.1 | - | 5 - 6 | 1091 | 57 |