Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina5
Entrez Gene ID
268591
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 5
Gene Aliases
4933415L04, PAI-3, Pci
Chromosome Location
Chr.12: 104099313 - 104106138 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172953.3 NP_766541.2 2 - 3 777 81
GenBank mRNA AK087714.1 - 2 - 3 740 81
BC090981.1 - 2 - 3 740 81

View Full Product Details