The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008865.3 | NP_032891.1 | 3 - 4 | 365 | 79 |
GenBank mRNA | AK005542.1 | - | 3 - 4 | 366 | 79 |
AK005543.1 | - | 3 - 4 | 365 | 79 | |
AK131667.1 | - | 3 - 4 | 366 | 79 | |
AK167483.1 | - | 3 - 4 | 366 | 79 | |
BC099386.1 | - | 3 - 4 | 350 | 79 | |
M14647.1 | - | 3 - 4 | 365 | 79 |