The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008937.2 | NP_032963.1 | 4 - 5 | 2509 | 64 |
GenBank mRNA | AF061576.1 | - | 3 - 4 | 2096 | 64 |
AK052889.1 | - | 4 - 5 | 2543 | 64 | |
AK143179.1 | - | 4 - 5 | 2333 | 64 | |
AK218220.1 | - | 1 - 2 | 203 | 64 | |
BC051411.1 | - | 4 - 5 | 2509 | 64 |