Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptprn
Entrez Gene ID
19275
Gene Name
protein tyrosine phosphatase, receptor type, N
Gene Aliases
IA-2, mIA-A, mKIAA4064
Chromosome Location
Chr.1: 75247035 - 75264497 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008985.2 NP_033011.2 7 - 8 1209 54
GenBank mRNA AF332083.1 - 4 - 5 813 54
AF332084.1 - 4 - 5 813 54
AK041296.1 - 7 - 8 1215 54
AK076223.1 - 7 - 8 1190 54
AK139398.1 - 6 - 7 1729 54
AK163141.1 - 7 - 8 1187 54
AK220467.1 - 7 - 8 1167 54
BC068165.1 - 7 - 8 1141 54
DQ832283.1 - 7 - 8 1186 54
U11812.1 - 7 - 8 1127 54
X74438.1 - 7 - 8 1226 54

View Full Product Details