Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps6ka1
Entrez Gene ID
20111
Gene Name
ribosomal protein S6 kinase polypeptide 1
Gene Aliases
Mapkapk-1a, Rsk, Rsk-1, Rsk1, S6K-alpha-1, p90-Rsk1, p90Rsk1, p90S6K, p90rsk
Chromosome Location
Chr.4: 133847290 - 133887797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285505.1 NP_001272434.1 12 - 13 1117 111
NM_001285506.1 NP_001272435.1 11 - 12 1001 111
NM_009097.5 NP_033123.2 12 - 13 1150 111
GenBank mRNA AK132856.1 - 10 - 11 1187 111
AK148208.1 - 12 - 13 1143 111
AK167790.1 - 11 - 12 988 111
BC049076.1 - 12 - 13 1101 111
BC094470.1 - 12 - 13 1103 111
M28489.1 - 12 - 13 1103 111

View Full Product Details