The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009142.3 | NP_033168.2 | 1 - 2 | 151 | 72 |
GenBank mRNA | AF010586.1 | - | 1 - 2 | 105 | 72 |
AF071549.1 | - | 1 - 2 | 142 | 72 | |
AK040220.1 | - | 1 - 2 | 96 | 72 | |
AK078285.1 | - | 1 - 2 | 128 | 72 | |
AK138214.1 | - | 1 - 2 | 128 | 72 | |
AK138472.1 | - | 1 - 2 | 128 | 72 | |
AK138574.1 | - | 1 - 2 | 128 | 72 | |
AK154646.1 | - | 1 - 2 | 128 | 72 | |
BC006650.1 | - | 1 - 2 | 105 | 72 | |
BC054838.1 | - | 1 - 2 | 111 | 72 | |
CB526167.1 | - | 1 - 2 | 151 | 72 | |
U92565.1 | - | 1 - 2 | 66 | 72 |