The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243072.1 | NP_001230001.1 | 4 - 5 | 715 | 67 |
NM_001243073.1 | NP_001230002.1 | 4 - 5 | 555 | 67 | |
NM_009152.4 | NP_033178.2 | 3 - 4 | 983 | 67 | |
GenBank mRNA | AK053115.1 | - | 3 - 4 | 716 | 67 |
AK140817.1 | - | 4 - 5 | 556 | 67 | |
AK164242.1 | - | 6 - 7 | 758 | 67 | |
BC057588.1 | - | 3 - 4 | 630 | 67 | |
BC090844.1 | - | 4 - 5 | 715 | 67 | |
D85028.2 | - | 3 - 4 | 985 | 67 | |
L41541.1 | - | 3 - 4 | 334 | 67 | |
X85993.1 | - | 3 - 4 | 444 | 67 |