The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009374.3 | NP_033400.2 | 12 - 13 | 1999 | 110 |
GenBank mRNA | AK036783.1 | - | 3 - 4 | 445 | 110 |
BC129890.1 | - | 12 - 13 | 1965 | 110 | |
BC129891.1 | - | 12 - 13 | 1965 | 110 | |
L10385.1 | - | 12 - 13 | 1989 | 110 |