The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165894.1 | NP_001159366.1 | 9 - 10 | 1204 | 64 |
NM_009652.3 | NP_033782.1 | 9 - 10 | 1204 | 64 | |
GenBank mRNA | AK040758.1 | - | 9 - 10 | 1010 | 64 |
AK154936.1 | - | 9 - 10 | 985 | 64 | |
BC066018.1 | - | 9 - 10 | 1204 | 64 | |
M94335.1 | - | 8 - 9 | 856 | 64 | |
X65687.1 | - | 9 - 10 | 1117 | 64 |