The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036684.2 | NP_001031761.1 | 2 - 3 | 635 | 97 |
NM_009723.5 | NP_033853.1 | 4 - 5 | 824 | 97 | |
GenBank mRNA | AB030737.1 | - | 1 - 2 | 196 | 97 |
AB030738.1 | - | 1 - 2 | 196 | 97 | |
AF053471.1 | - | 3 - 4 | 653 | 97 | |
AK051287.1 | - | 2 - 3 | 630 | 97 | |
AK054112.1 | - | 3 - 4 | 786 | 97 | |
AK147240.1 | - | 4 - 5 | 710 | 97 | |
AK147357.1 | - | 2 - 3 | 564 | 97 | |
AK147369.1 | - | 4 - 5 | 760 | 97 | |
AK147399.1 | - | 2 - 3 | 799 | 97 | |
AK147501.1 | - | 4 - 5 | 771 | 97 | |
BC075643.1 | - | 2 - 3 | 635 | 97 | |
GU734816.2 | - | 1 - 2 | 196 | 97 |