The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037134.2 | NP_001032211.1 | 6 - 7 | 670 | 88 |
NM_001282943.1 | NP_001269872.1 | 6 - 7 | 583 | 88 | |
NM_009830.3 | NP_033960.1 | 6 - 7 | 580 | 88 | |
GenBank mRNA | AF091432.1 | - | 5 - 6 | 475 | 88 |
AF106691.1 | - | 4 - 5 | 450 | 88 | |
AK010929.1 | - | 6 - 7 | 510 | 88 | |
AK011883.1 | - | 1 - 2 | 142 | 88 | |
AK031116.1 | - | 6 - 7 | 507 | 88 | |
AK037791.1 | - | 6 - 7 | 510 | 88 | |
AK158720.1 | - | 6 - 7 | 1096 | 88 | |
AK167428.1 | - | 6 - 7 | 510 | 88 | |
BC053727.1 | - | 5 - 6 | 470 | 88 |