The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010071.2 | NP_034201.1 | 3 - 4 | 498 | 79 |
GenBank mRNA | AF030627.1 | - | 3 - 4 | 498 | 79 |
AF035117.1 | - | 3 - 4 | 494 | 79 | |
AF059583.1 | - | 3 - 4 | 484 | 79 | |
AK155579.1 | - | 3 - 4 | 495 | 79 | |
AK158246.1 | - | 3 - 4 | 462 | 79 | |
AK159466.1 | - | 3 - 4 | 539 | 79 | |
AK159472.1 | - | 3 - 4 | 576 | 79 | |
BC004590.1 | - | 3 - 4 | 490 | 79 |