The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010149.3 | NP_034279.3 | 6 - 7 | 972 | 70 |
GenBank mRNA | AK139031.1 | - | 6 - 7 | 871 | 70 |
BC003953.1 | - | 6 - 7 | 825 | 70 | |
J04843.1 | - | 6 - 7 | 854 | 70 |