The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010149.3 | NP_034279.3 | 7 - 8 | 1054 | 70 |
GenBank mRNA | AK010968.1 | - | 6 - 7 | 762 | 70 |
AK139031.1 | - | 7 - 8 | 953 | 70 | |
BC003953.1 | - | 7 - 8 | 907 | 70 | |
BC046282.1 | - | 7 - 8 | 984 | 70 | |
J04843.1 | - | 7 - 8 | 936 | 70 |