The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008607.2 | NP_032633.1 | 8 - 9 | 1254 | 149 |
GenBank mRNA | AK150728.1 | - | 8 - 9 | 1241 | 149 |
AK151626.1 | - | 8 - 9 | 1241 | 149 | |
AK152507.1 | - | 8 - 9 | 1240 | 149 | |
AK152916.1 | - | 8 - 9 | 1241 | 149 | |
AK153458.1 | - | 8 - 9 | 1239 | 149 | |
AK155188.1 | - | 8 - 9 | 1240 | 149 | |
AK159921.1 | - | 8 - 9 | 1241 | 149 | |
AK160819.1 | - | 8 - 9 | 1239 | 149 | |
BC125320.1 | - | 8 - 9 | 1254 | 149 | |
BC125322.1 | - | 8 - 9 | 1254 | 149 | |
X66473.1 | - | 8 - 9 | 1223 | 149 |