Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il6st
Entrez Gene ID
16195
Gene Name
interleukin 6 signal transducer
Gene Aliases
5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130
Chromosome Location
Chr.13: 112464070 - 112506860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010560.3 NP_034690.3 14 - 15 2116 89
GenBank mRNA AK142637.1 - 4 - 5 577 89
BC058679.1 - 14 - 15 2077 89
M83336.1 - 13 - 14 1921 89
X62646.1 - 13 - 14 1921 89

View Full Product Details